Learn More
Medchemexpress LLC MMP-9-IN-6 | 2241964-36-1 | 99.8% | 365.42 | 25 MG

Supplier: Medchemexpress LLC 25MGHY149848
MMP-9-IN-6 (Compound 3g) is an MMP-9 inhibitor with an IC50 value of 50 μM, demonstrating a good anti-ulcer effect. This product is for research use only and is not sold to patients.
- MMP-9 inhibitor
- IC50 value of 50 μM
- Demonstrates good anti-ulcer effect
- Purity of 99.8%
- Solid appearance
- Color is pale purple to purple
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.