Learn More
Medchemexpress LLC MMP-9-IN-9 10mg | 206549-55-5 | 511.63 | C27H33N3O5S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11630210MG
Encompass_Preferred
MMP-9-IN-9 is a selective small-molecule inhibitor of matrix metalloproteinase-9 (MMP-9) for research use. It exhibits potent inhibition of MMP-9 (IC50 ≈ 5 nM), shows selectivity versus other MMPs, is supplied as a white to off-white solid, and is typically dissolved in DMSO for in vitro studies. Recommended storage for the powder is -20°C.
Catalog No. 50-003-08284