Learn More
Medchemexpress LLC MMP3 inhibitor 3 TFA | 686.78 (free base) | 10 MG

Supplier: Medchemexpress LLC HYP2166A10MG
MMP3 inhibitor 3 (TFA) is an inhibitor of MMP3. It can be used to study breast cancer. It has the molecular formula C27H46N10O9S.XC2HF3O2 and a molecular weight of 686.78 (free base). The sequence is Ac-Arg-Cys-Gly-Val-Pro-Asp-NH2, shortened to Ac-RCGVPD-NH2. Its target is MMP, and the pathway is Metabolic Enzyme/Protease.
- Inhibits MMP3
- Can be used to study breast cancer
- Sealed storage, away from moisture
- Powder is stable for 2 years at -80°C
- Powder is stable for 1 year at -20°C
- In solvent, it is stable for 6 months at -80°C
- In solvent, it is stable for 1 month at -20°C
- Soluble in DMSO at 100 mg/mL (needs ultrasonic)
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.