Learn More
ABclonal Technology MMP3 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2232820UL
Encompass_Preferred
Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes such as embryonic development reproduction and tissue remodeling as well as in disease processes such as arthritis and metastasis Most MMP s are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases This gene encodes an enzyme which degrades fibronectin laminin collagens III IV IX and X and cartilage proteoglycans The enzyme is thought to be involved in wound repair progression of atherosclerosis and tumor initiation The gene is part of a cluster of MMP genes which localize to chromosome 11q22 3
Catalog No. 50-000-05816