Learn More
Medchemexpress LLC HY-19351 5mg Medchemexpress, MN-64 CAS:92831-11-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY193515MG
Encompass_Preferred
Medchemexpress, HY-19351 5mg MN-64 CAS:92831-11-3 MN-64 is a potent tankyrase 1 inhibitor, with IC50s of 6 nM, 72 nM, 19.1 μM, and 39.4 μM for TNKS1, TNKS2, ARTD1 and ARTD2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2124