Learn More
Medchemexpress LLC Mn(II)-DO3A sodium | 120066-53-7 | 70.0% | 421.28 | C14H23MnN4NaO6 | 10 MG

Supplier: Medchemexpress LLC HY15733510MG
Mn(II)-DO3A sodium is a manganese(II) complex supplied as its sodium salt for research into MRI contrast agents and as a potential alternative to gadolinium-based agents. It is provided as a solid reagent intended for laboratory research use only.
- Mn(II) complex designed as alternative to gadolinium-based MRI contrast agents.
- Provided as the sodium salt for improved solubility and handling.
- Available in small research quantities (e.g., 10 mg) for laboratory studies.
- Reported purity approximately 70.0%.
- Molecular weight 421.28 and formula C14H23MnN4NaO6.
- CAS number 120066-53-7 for unambiguous identification.
- Documentation available: data sheet, SDS, and COA for handling and characterization.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.