Learn More
Enzo Life Sciences LAG-3 (human) monoclonal antibody (17B4) (ATTO 647N conjugate) (100 tests)
SDP
Supplier:  Enzo Life Sciences ALX804806TST100
Encompass_Preferred
Clone: 17B4. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to 30 aa (GPPAAAPGHPLAPGPHPAAPSSWGPRPRRY) from the first N-terminal D1 domain of human LAG-3 (lymphocyte activation gene-3). UniProt: P18627. Source: Purified from hybridoma tissue culture supernatant. Species Reactivity: Human. Specificity: Recognizes the 30 aa extra-loop of the first N-terminal D1 domain of LAG-3. Applications: Flow Cytometry, ICC. Recommended dilutions: Flow Cytometry (1:100). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Purified from concentrated hybridoma tissue culture supernatant by Protein A Sepharose™ CL-4B Flow Fast Column. Formulation: Liquid. In PBS containing 50mM Tris, 1% BSA and 0.02% sodium azide. Handling: Avoid vigorous centrifugation and vortexing. Do not freeze. Protect from light.
Catalog No. NC1696346