Learn More
Proteintech Group Inc mNeonGreen Monoclonal Antibody, Clone 32F6, Unconjugated, 100 μL, Protein A purified
SDP
Supplier:  Proteintech Group Inc 32F6100
Encompass
The mNeonGreen Monoclonal Antibody is a highly specific mouse monoclonal antibody designed to detect mNeonGreen fluorescent protein derived from *Branchiostoma lanceolatum*. This purified unconjugated antibody is suitable for a variety of applications including immunofluorescence, Western Blot, and ELISA. It offers robust performance with minimal cross-reactivity, making it a reliable choice for researchers working with mNeonGreen-tagged fusion proteins. The antibody is stored in a buffered solution and is research-ready, ensuring high consistency and reproducibility in experimental results.
  • High specificity to mNeonGreen without cross-reactivity to EGFP
  • Validated for use in immunofluorescence, Western Blot, and ELISA
  • Reliable monoclonal format ensuring consistent results
  • Reacts with mNeonGreen at N-terminal, C-terminal, or internal fusion sites
  • Supplied in a buffered aqueous solution, ready for research applications
Catalog No. NC2047613