Learn More
Cayman Chemical MnIdIpIn-d4 1mg
SDP
Supplier:  Cayman Chemical 307681 MG
Encompass_Preferred
An internal standard for the quantification of manidipine by GC- or LC-MS
Catalog No. 50-298-8857