Learn More
Biotium Mix-n-Stain™ DNP Antibody Labeling Kit, 1x(20-50ug) labeling
Supplier:  Biotium 92326
Encompass_Preferred
Mix-n-Stain™ DNP antibody labeling kits are a revolutionary antibody labeling technology that allows you to label your antibody with DNP in 30 minutes without a purification step. The labeling procedure tolerates many common buffer components and antibody stabilizers. Choose from kit sizes to label 5-20 ug, 20-50 ug, or 50-100 ug of your antibody.
Catalog No. 50-196-4977