Learn More
Enzo Life Sciences MnTBAP chloride (50mg)
SDP
Supplier:  Enzo Life Sciences ALX430069M050
Encompass_Preferred
Cell permeable superoxide dismutase (SOD) mimetic. Potent inhibitor of peroxynitrite-induced oxidative reactions (peroxynitrite scavenger), but not a scavenger of nitric oxide (NO). Inhibits neuronal apoptosis. Purity: ≥95% (TLC). Solubility: Dissolve at 0.1M in aqueous base, then add buffer to lower pH to pH 7.0 or higher as needed. Long Term Storage: -20°C.
Catalog No. 50-200-9178