Learn More
Medchemexpress LLC MnTBAP (chloride) | 55266-18-7 | 98.0% | 879.15 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12639710MM/1ML
Encompass_Preferred
MnTBAP chloride is a manganic porphyrin complex that acts as a superoxide dismutase (SOD) mimetic and peroxynitrite scavenger, demonstrating anti-oxidative properties. It mediates anti-inflammatory effects by upregulating BMPR-II and inhibiting NFκB signaling, showing potential for research into renal fibrosis and chronic kidney diseases.
  • Mediates anti-inflammatory effects through upregulation of BMPR-II.
  • Inhibits NFκB signaling.
  • Exhibits anti-oxidative properties.
  • Functions as a superoxide dismutase mimetic.
  • Acts as a peroxynitrite scavenger.
  • Shows potential in attenuating renal fibrosis.
  • Suppresses α-SMA and vimentin induction.
  • Increases BMPR-II mRNA expression.
Catalog No. 50-004-36239