Learn More
Medchemexpress LLC MnTE-2-PyP chloride | 219818-60-7 | 95.0% | C48H44Cl5MnN8 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13057410MG
Encompass_Preferred
MnTE-2-PyP chloride is a manganese porphyrin compound for research use that functions as a reactive oxygen species (ROS) scavenger and a potent radioprotector. Supplied as a brown-to-black solid with approximately 95.0% purity, it is intended for studies of oxidative stress, tissue radioprotection, and related biological applications.
  • Reactive oxygen species scavenger and radioprotector.
  • Manganese porphyrin structure provides redox activity.
  • Supplied as a brown-to-black solid with 95.0% purity.
  • Storage recommendations provided for solid and solvent conditions.
  • Suitable for in vitro and in vivo oxidative stress and radioprotection research.
  • For research use only.
Catalog No. 50-002-60661