Learn More
Medchemexpress LLC Mo-i-1100 | 1585217-40-8 | 99.8% | 379.81 g/mol | C17H14ClNO5S | 10 MG

Supplier: Medchemexpress LLC HY16441010MG
MO-I-1100 is a small-molecule inhibitor of aspartate beta-hydroxylase (ASPH) supplied for in vitro and preclinical research. It has reported antitumor activity via inhibition of Notch signaling and suppresses hepatocellular carcinoma cell migration, invasion, and anchorage-independent growth. The compound is provided as a solid with accompanying documentation for handling and analysis.
- Small-molecule ASPH inhibitor for research use only.
- Molecular weight 379.81 g/mol; formula C17H14ClNO5S.
- CAS number 1585217-40-8.
- Purity approximately 99.8%.
- Appearance white to off-white solid.
- Available in small package sizes (for example, 10 mg) with SDS and COA available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.