Learn More
Medchemexpress LLC MO-I-500 | 1585219-04-0 | 99.1% | 317.75 g/mol | C12H12ClNO5S | 10 MG

Supplier: Medchemexpress LLC HY11742610MG
MO-I-500 is a small-molecule research inhibitor of the FTO demethylase. It inhibits purified FTO with an IC50 of 8.7 μM and is supplied as a white to off-white solid for in vitro laboratory use.
- Inhibits FTO demethylase (IC50 8.7 μM).
- High purity (99.1%).
- Molecular weight 317.75 g/mol; formula C12H12ClNO5S.
- Soluble in DMSO (31.8 mg/mL); ultrasonic and warming recommended.
- Powder stable at -20°C for up to 3 years.
- Provided as a 10 mg solid for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.