Learn More
Innovative Research Inc moAbs M5114 PE/Cyin 5.5
Supplier:  Innovative Research Inc IRTAMSMHCIIM5114CPECY551
Encompass_Preferred
Anti Mouse MHC II (I-A/I-E) Flow Cytometry Monoclonal Clone M5114 PE/Cyanine5.5 100ug - Anti Mouse MHC II (I-A/I-E) Flow Cytometry Monoclonal Clone M5114 PE/Cyanine5.5 from Innovative Research is a flow cytometry antibody, buffered in PBS with 0.05% Proclin300, 1% BSA. This flow cytometry antibody has been specifically designed to work with our flow cytometry isotype controls. Our scientists suggest selection first be made of your desired flow cytometry antibody as these product descriptions contain direct links to relevant isotype controls. Choose between flow cytometry antibodies offered by mass, which are ready for you to dilute to your specific titration requirements, or our ready to use format, which have been developed for ease of use under typical assay conditions as described in the product documentation. RUO
Catalog No. 50-274-4014