Learn More
Seca Corp 210 Mobile vinyl measuring mat for babies and toddlers, in/cm
SDP
Supplier:  Seca Corp 2101821004
Encompass_Preferred
The seca 210 is a lightweight and space-saving solution for precise length measurement of babies and toddlers while lying down. While kind to the skin, the 210 is washable and foldable. The fixed head piece and the sliding foot positioner make it simple to use. • Measuring range 4 – 39 inches / 10 – 99 cm • Graduation 1/4 inch / 5 mm • Accessories: Backpack seca 409 or carrying case seca 414 (for combination with baby scale seca 354)
Catalog No. NC1798758