Learn More
Mascon Medical MobiMicrobe® High Throughput Microbial Single-Cell DNA-seq Kit, 4rxn
SDP
Supplier:  Mascon Medical MMMSDNA4
Encompass_Preferred
Consumables and reagents used in conjunction with the MobiNova-M1. This kit is suitable for Gram negative/positive bacteria, and the experiment includes multiple processes such as microbial lysis, whole genome amplification, gene fragmentation, and library construction. It can be used to detect the full-length genome of a single microorganism.
Catalog No. NC3802111
Add to Cart
Request a Quote