Learn More
Mascon Medical MobiNova®-D1 Tissue Dissociation Instrument
SDP
Supplier:  Mascon Medical MMD1TDI
Encompass_Preferred
"MobiNova-D1 Tissue Dissociation Instrument combines the principles of mechanical fragmentation and enzyme digestion to dissociate animal tissues, and can obtain high activity, high yield, and high repeatability single-cell suspensions or tissue homogenates in a short period of time."
Catalog No. NC3802101
Only null left
Add to Cart
Request a Quote