Learn More
Medchemexpress LLC HY-135815A 10mg Medchemexpress, Mobocertinib (succinate) CAS:2389149-74-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135815A10MG
Encompass_Preferred
Medchemexpress, HY-135815A 10mg Mobocertinib (succinate) CAS:2389149-74-8 Mobocertinib succinate (TAK-788 succinate) is a potent and orally active inhibitor of EGFR and HER2 oncogenic mutants, including exon 20 insertions, with selectivity over WT EGFR. Antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1404