Learn More
Selleck Chemical LLC Mocetinostat (MGCD0103) 10mM/1mL 726169-73-9 MG0103
SDP
Supplier:  Selleck Chemical LLC S112210MM/1ML
Encompass_Preferred
Mocetinostat (MGCD0103, MG0103) is a potent HDAC inhibitor with most potency for HDAC1 with IC50 of 0.15 ?M in a cell-free assay, 2- to 10- fold selectivity against HDAC2, 3, and 11, and no activity to HDAC4, 5, 6, 7, and 8. Mocetinostat (MGCD0103) induces apoptosis and autophagy. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-0938