Learn More
Cygnus Technologies Cygnus Technologies MockV RVLP Kit, Economical Retroviral Clearance, BSL-1 Compatible Stock Solution, 2.0 mL
Supplier:  Cygnus Technologies M230
Encompass
The Cygnus Technologies MockV RVLP Kit is designed for the economical and timely quantification of retrovirus-like particle clearance in biopharmaceutical manufacturing. This kit allows researchers to conduct viral clearance studies in-house, avoiding expensive CRO studies with live viruses. It includes BSL-1 compatible RVLP stock solution from CHO cells and provides sensitive detection and accurate quantification, streamlining the development process and ensuring purification efficacy early on.
Catalog No. NC2445707