Learn More
YSI 3P LifeScience Model 3252 Conductivity Probe
Supplier:  YSI 3P LifeScience 3252
Encompass_Preferred
YSI 3200 series conductivity cells have built-in thermistors, allowing automatic temperature compensation. All YSI cells are calibrated according to the International Organization of Legal Metrology recommendations 56 and 68. Shipped with greater than 1% cell accuracy; includes Certificate of Traceability. Can be used as secondary lab standard. Cells can be re-calibrated (adjusted) to NIST traceable standards; YSI also offers this service with a Certificate of Calibration and Traceability if needed. Black platinum electrodes are extremely stable and linear; can be replatinized using the YSI 3100 or 3200 instruments and 3140 platinizing solution
Catalog No. NC1280901
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.