Learn More
Electron Microscopy Sciences ThermoSafe Model 339 LabMailer
SDP
Supplier:  Electron Microscopy Sciences 6151539
Encompass_Preferred
Maximum breakage protection for known non-hazadous blood, serum, tissue, and other specimens. Accommodates  VacuTainer  Microtainer and other collection / transport tubes. Lightweight polystyrene foam construction saves postage. Reusable and recyclable. Holds 5 tubes, 105 x 13ml, 365 & 340 Mailing Sleeve.
Catalog No. 50-338-03