Learn More
Sentry Air Systems Model 400 Acid Gas Filter
SDP
Supplier:  Sentry Air Systems SS422AGF
Encompass
Model 400/450 Acid Gas Filter features 2.5 lbs of heavy-duty activated carbon treated for acid gas removal. Fits Model 400 or 450 systems.
Catalog No. NC0450357