Learn More
Electron Microscopy Sciences U-tek Model 402 Mini Mailers
SDP
Supplier:  Electron Microscopy Sciences 6152202
Encompass_Preferred
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants. Insulating, thick wall polystyrene foam construction. Holds bottles, cans, vials, cups. Can be fitted with foam insert to hold small kit components.
Catalog No. 50-338-31