Learn More
Electron Microscopy Sciences U-tek Model 404 Mini Mailers
SDP
Supplier:  Electron Microscopy Sciences 6152204
Encompass_Preferred
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants. Insulating, thick wall polystyrene foam construction. Holds 10 standard petri dishes.
Catalog No. 50-338-26