Learn More
Electron Microscopy Sciences ThermoSafe Insulated Shipper Model 413, EPS Box with Carton, 11 x 9", 10" height

Supplier: Electron Microscopy Sciences 6153641
These shippers and containers offer the ideal solution for the safe storage and delivery of most refrigerated or frozen materials, including pharmaceuticals, laboratory specimens, serum and vaccines, human organs, culture packs, and nuclear and isotope materials. When shipping dry ice or utilizing dry ice as a refrigerant, these shippers provide excellent dry ice sublimation - average dry ice usage in 1 1/2 thick wall units is 5 lbs for the first 24 hours; 3 – 4 lbs thereafter.
Type: EPS Box with Carton
Size: 11 x 9"
Height: 10"
Thickness: 1.5"
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.