Learn More
Electron Microscopy Sciences ThermoSafe Model 436 Aluminum Can Shippers
SDP
Supplier:  Electron Microscopy Sciences 6151736
Encompass_Preferred
Accommodates the  most widely used laboratory collection tubes, vials, and bottles. Helps to prevent breakage during handling and shipment. Seamless construction with lined screw-cap to prevent leakage. completely autoclavable. EPS foam mailers and mailing sleeves available.
Diameter: 2.5"
OAL: 6"
Catalog No. 50-338-15