Learn More
Electron Microscopy Sciences U-tek Model 440 Mini Mailers for Temperature Sensitive Shipments
SDP
Supplier:  Electron Microscopy Sciences 6152040
Encompass_Preferred
Compact protection for temperature sensitive shipments, ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants. Insulating, thick wall polystyrene foam construction.
Catalog No. 50-338-29