Learn More
Electron Microscopy Sciences U-tek Model 602 Mailing Sleeves
SDP
Supplier:  Electron Microscopy Sciences 6152302
Encompass_Preferred
Corrugated Carton provides protection for temperature sensitive shipments. Ideal for serums, tissues, and reagents. Accommodate tubes, vials, ampoules, small bottles, and tissue flasks with space for refrigerants.
Catalog No. 50-338-32