Learn More
Electron Microscopy Sciences ThermoSafe Infectious Shipper Model 670
SDP
Supplier:  Electron Microscopy Sciences 6153570
Encompass_Preferred
These shippers are for shipping infectious agents in accordance with DOT Regulation 49CFR Parts 171 to 178 and IATA 602. They Meet US Dept of Transportation (DOT) and IATA requirement for transporting infectious substances (UN2814 and UN 2900).
Note: Compliance with UN/DOT regulations governing shipments of infectious substances in these approved shippers is contingent on the use of all supplied components. The shipper is ultimately responsible for determining the suitability of packaging for use with contents. Consult appropriate regulations. The EPS foam container, lid for Model 630 or Model 660B shipper plus 25 lbs of dry ice to protect frozen samples for four days.
Catalog No. 50-339-28