Learn More
Electron Microscopy Sciences Model 865 Diamond Band Saw
SDP
Supplier:  Electron Microscopy Sciences 5018265
Encompass
Constructed from a high impact polymer, the lightweight Model 865 Diamond Band Saw is designed for medium precision, with efficient cutting of hard and brittle materials. Rugged enough to cut through materials such as soft metals and polymers as well. Samples can be safely hand fed into the blade with or without the standard specimen guide. The diamond plated steel core blades are designed to last long under normal conditions. Easy blade access and replacement make occasional saw maintenance extremely simple. Coolant is contained in a reservoir at the base of the saw.
Small, lab bench size
Quick adjustable blade guide keeps the blade tracking properly.
Rugged, corrosion-proof construction.
Powerful motor cuts fast, won't bog down under load.
Minimum coolant splash for clean cutting.
Easy access coolant tray for fast and easy clean up.
Large table for cutting bulk samples.
Sample guide for straight, flat surfaces.
Catalog No. 50-997-136