Learn More
Electron Microscopy Sciences ThermoSafe Medical Transporter Model 931
SDP
Supplier:  Electron Microscopy Sciences 6153731
Encompass_Preferred
Insulated expanded polypropylene medical transporter is ideal for on-site collection, storage, and transport of temperature sensitive specimens, medications, medical devices and organs. Durable nylon clad transporter with impact resistant expanded polyethylene container. Lightweight ergonomic design. Extra space for crushed ice and refrigerant gel pack. Washable and weather resistant. Removable, rigid, thermoformed tray. Prevents leaking. 3/8 wall thickness.
Catalog No. 50-339-50