Learn More
Electron Microscopy Sciences ThermoSafe Model 954 Transport Bag
SDP
Supplier:  Electron Microscopy Sciences 6152554
Encompass_Preferred
Specimen transport bags are liquid tight and tamper evident. The patented adhesive closure and continuous seal meet or exceed standards of all major air carriers and DOT. Polyethylene construction and adhesive closure provides added security during handling by personnel.
I.D.: 13-1/4 x 17-7/8"
Catalog No. 50-339-08