Learn More
Electron Microscopy Sciences ThermoSafe Medical Transporter Model 960
SDP
Supplier:  Electron Microscopy Sciences 6153760
Encompass_Preferred
Insulated expanded polypropylene medical transporter is ideal for on-site collection, storage, and transport of temperature sensitive specimens, medications, medical devices and organs. Durable nylon clad transporter with impact resistant expanded polyethylene container. Lightweight ergonomic design. Extra space for crushed ice and refrigerant gel pack. Washable and weather resistant. Cools and stores whole blood units. 1 wall thickness.
Catalog No. 50-339-52