Learn More
Alkali Scientific Alkali Scientific™ Modifiable Upright Freezer Rack for Standard 2" or 3" Boxes, 4x6 Configuration
SDP
Supplier:  Alkali Scientific UFMC4623
Encompass
Modifiable Upright Freezer Rack Our Freezer Storage Racks are constructed of high-quality, corrosion resistant stainless steel. Handles at both ends accommodate freezers with right or left handed doors.  Our freezer racks have been configured to fit all major ultra low (-80°) freezers on the market; racks are designed for ultra low upright freezers.  Removable clip can be adjusted so that the rack can hold both standard 2" and 3" boxes.Constructed of corrosion resistant stainless steel. Features: Modifiable Includes Handles Constructed of corrosion resistant stainless steel Dimensions (DxWxH): 22 3/8 x 5 1/2 x 12 5/8inches (569 x 140 x 321mm)Compartment Dimensions (2″ Boxes) L x H x W: 5 7/16  x 2 3/16 x 5 3/8 Inches  (138 x 54.6 x 137mm)Compartment Dimensions (3″ Boxes) L x H x W: 5 7/16 x 3 1/16 x 5 3/8 inches  (138 x 77.7 x 137m)
Catalog No. 50-302-9485