Learn More
Teknova Modified 2X M9 Salts Media , 500mL Sterile.
SDP
Supplier:  Teknova M2950
Encompass
M9 minimal media is a chemically-defined growth medium that was developed for use with Escherichia coli. It is a highly versatile medium that can be used for a variety of purposes, including isolating and culturing bacteria, performing genetic transformation experiments, and growing bacteria for use in bioreactors.
Catalog No. 50-238-8030