Learn More
Sino Biological Peptide Substrates: Modified AKT Substrate II
Supplier:  Sino Biological A0558C
Encompass_Preferred
The Modified AKT Substrate II peptide sequence (modified-CKRPRAASFAE) is based on the N-terminus of GSK3.
Catalog No. 50-256-9865