Learn More
Sino Biological Peptide Substrates: Modified AXLtide Substrate
Supplier:  Sino Biological A1658B
Encompass_Preferred
The Axltide peptide sequence (CKKSRGDYMTMQIG) is based on the mouse Insulin receptor substrate 1 (amino acid 979-989).
Catalog No. 50-256-9823