Learn More
Sino Biological Peptide Substrates: Modified SGKtide
Supplier:  Sino Biological S0858B
Encompass_Preferred
The Modified SGKtide peptide sequence (Modified- CKKRNRRLSVA) contains serine protein kinase phosphorylation site and is evaluated as a substrate for SGK and NDR family kinases.
Catalog No. 50-256-7890