Learn More
Medchemexpress LLC Modoflaner | 1331922-53-2 | 98.26% | 715.23 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY137445100MG
Encompass_Preferred
Modoflaner is an isophenylamide insecticide that may act through allosteric regulation of gamma-aminobutyric acid-gated chloride channels. It is intended for research use only.
  • Kills more than 70% of the moth, xylophone, and gray planthopper in vitro
  • Prevents female adult ticks from laying eggs or eggs from hatching in vivo
Catalog No. 50-003-50193