Learn More
New England Biolabs, Inc. Trypsin-ultra™, Mass Spectrometry Grade – 100 µg
SDP
Supplier:  New England Biolabs, Inc. P8101S
Encompass
Trypsin-ultra, Mass Spectrometry Grade is a serine endopeptidase, which selectively cleaves peptide bonds C-terminal to lysine and arginine residues. Trypsin-ultra cleaves at Lys-Pro and Arg-Pro bonds at a much slower rate than when Lys and Arg are N-terminal to other residues.
Catalog No. 50-811-916