Learn More
Electron Microscopy Sciences Dri-Bath™ Modular Block for Machining by user, 2078Q
SDP
Supplier:  Electron Microscopy Sciences 6313210
Encompass_Preferred
Close contact of tubes to block walls, provides excellent heat transfer. Block includes thermometer well for monitoring block temperatures. Solid black anodized aluminum blocks for better heat retention. Block Puller included with all models. For use with all our brand modular dri-baths.
Model: 2078Q
Catalog No. 50-322-80