Learn More
Cayman Chemical MoexIprIl-d5 10mg
SDP
Supplier:  Cayman Chemical 3381410 MG
Encompass_Preferred
An internal standard for the quantification of moexipril by GC- or LC-MS
Catalog No. 50-294-8705