Learn More
Cayman Chemical Moexprlat 5mg
SDP
Supplier:  Cayman Chemical 380475 MG
Encompass_Preferred
An ACE inhibitor and active metabolite of the prodrug moexipril (IC50 = 2.1 nM); prevents the estrone- or angiotensin II-stimulated proliferation of primary neonatal rat cardiac fibroblasts at 10 nM; reduces mean arterial blood pressure and increases the levels of atrial natriuretic peptide in ovariectomized mice at 50 mg/kg per day
Catalog No. 50-291-8034