Learn More
Medchemexpress LLC Mofezolac | 78967-07-4 | 99.3% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY120824100MG
Encompass_Preferred
Mofezolac is a non-steroidal anti-inflammatory research compound and a selective, reversible COX-1 inhibitor used in studies of inflammation and pain pathways. It is supplied in laboratory quantities and characterized for molecular properties and purity.
  • Selective, reversible COX-1 inhibitor suitable for inflammation research.
  • High chemical purity for analytical and experimental reproducibility.
  • Well-characterized molecular weight and formula for formulation and dosing.
  • Available in multiple small pack sizes to support screening and assay workflows.
  • COA and SDS available to support safe handling and quality assurance.
Catalog No. 50-002-33688