Learn More
Medchemexpress LLC Mog (35-55) (acetate) | 149635-73-4 | 99.7% | 2581.95 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP1240
Encompass_Preferred
MOG (35-55) mouse, rat, a minor component of CNS myelin, exhibits encephalitogenic activity and stimulates T cell proliferation. It induces a Th1 cytokine response and high levels of IgG antibodies, leading to a relapsing-remitting neurological disease with extensive plaque-like demyelination. This product is suitable for experimental autoimmune encephalomyelitis (EAE) modeling.
  • Exhibits encephalitogenic activity
  • Stimulates T cell proliferation
  • Induces Th1 cytokine response
  • Increases IgG levels in vitro
  • Useful for experimental autoimmune encephalomyelitis (EAE) modeling
  • Triggers immune response against the central nervous system
Catalog No. 50-004-26564