Learn More
Medchemexpress LLC MOG(35-55) amide, mouse, rat | 2022956-48-3 | >99.5% | 2580.97 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP371910MG
Encompass_Preferred
MOG (35-55) amide, mouse, rat is the terminal amidation form of the 35-55 fragment of the myelin oligodendrocyte glycoprotein (MOG) immunogenic peptide. It can be used for experimental autoimmune encephalomyelitis (EAE) modeling.
  • Can be used for experimental autoimmune encephalomyelitis (EAE) modeling.
  • Expanded CD4+ T cells are largely specific for the myelin oligodendrocyte glycoprotein (MOG) immunogenic peptide 35-55.
  • Can induce a classic EAE model characterized by myelin sheath damage and flaccid paralysis in ABH mice.
  • A known encephalitis-inducing epitope in C57BL/6 mice, capable of inducing chronic acute exacerbation encephalitis (EAE).
Catalog No. 50-003-97390