Learn More
Medchemexpress LLC MOG(35-55) amide, mouse, rat | 2022956-48-3 | 99.5% | 2580.97 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYP371950MG
Encompass_Preferred
MOG (35-55) amide, mouse, rat is the terminal amidation form of the 35-55 fragment of the myelin oligodendrocyte glycoprotein (MOG) immunogenic peptide. This product is suitable for use in experimental autoimmune encephalomyelitis (EAE) modeling.
  • Terminal amidation form of the 35-55 fragment of the myelin oligodendrocyte glycoprotein (MOG) immunogenic peptide.
  • Used for experimental autoimmune encephalomyelitis (EAE) modeling.
  • Expanded CD4+ T cells show specificity for this peptide.
  • Induces a classic EAE model in ABH mice, with myelin sheath damage and flaccid paralysis.
  • Recognized as an encephalitis-inducing epitope in C57BL/6 mice, capable of inducing chronic acute exacerbation EAE.
Catalog No. 50-003-99343